Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

Catalog No. 8615101G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-151-01G 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 86-151-01G Supplier Sigma Aldrich Supplier No. 8615101G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Korasilon is a trademark of kurt obermeier gmbh & co
  • Kg

Specifications

CAS 207738-08-7
Molecular Formula C16H20N2 -+ 2 HCl -+ xH2O
Linear Formula [-C6H2(CH3)2-4-NH2]2-+2 HCl-+xH2O
MDL Number MFCD00150104
Quantity 1 Ea.
Synonym TMB dihydrochloride
Molecular Weight (g/mol) 313.27 (anhydrous basis)
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.