Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetramethylbiphenyl, 97+%

Catalog No. aal0917203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0917203 1 g
AAL0917206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0917203 Supplier Thermo Scientific Chemicals Supplier No. L0917203
Only null left
Add to Cart

CAS: 25570-02-9 | C16H18 | 210.32 g/mol

3,3',5,5'-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3',5,5'-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3',5,5'-tetracarboxylic acid by the oxidation of with potassium permanganate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25570-02-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00051910
InChI Key CMZYGFLOKOQMKF-UHFFFAOYSA-N
PubChem CID 520212
IUPAC Name 1-(3,5-dimethylphenyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Melting Point 47°C to 49°C
Quantity 1 g
Beilstein 1935931
Synonym 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl #
Formula Weight 210.32
Percent Purity ≥97%
Chemical Name or Material 3,3',5,5'-Tetramethylbiphenyl
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.