Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3,5-bis(trifluoromethyl)phenoxy)-N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanam. | 1928724-23-5 | MFCD32174353 | 100.0% | C20H13F9N2O3 | 10MG
SDP

Catalog No. 5000258296
Encompass_Preferred

SK33 is a trifluoromethylated enobosarm analog research compound that acts as a tissue-selective anti-androgen by reducing androgen receptor (AR) transcriptional activity. It is used in biochemical and in vivo studies to probe nuclear receptor signaling and androgen-related pathways.

  • Tissue-selective anti-androgen activity that reduces AR transcriptional activity.
  • Target: androgen receptor; pathway: vitamin D related/nuclear receptor.
  • High purity (99.98%) suitable for in vitro assays.
  • Molecular formula C20H13F9N2O3; molecular weight 500.31.
  • Soluble in DMSO (100 mg/mL); stock solution and in vivo formulation guidance provided.
  • Storage recommendations: 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-58296 Supplier Medchemexpress LLC Supplier No. HY13573210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SK33 is a trifluoromethylated enobosarm analog research compound that acts as a tissue-selective anti-androgen by reducing androgen receptor (AR) transcriptional activity. It is used in biochemical and in vivo studies to probe nuclear receptor signaling and androgen-related pathways.

  • Tissue-selective anti-androgen activity that reduces AR transcriptional activity.
  • Target: androgen receptor; pathway: vitamin D related/nuclear receptor.
  • High purity (99.98%) suitable for in vitro assays.
  • Molecular formula C20H13F9N2O3; molecular weight 500.31.
  • Soluble in DMSO (100 mg/mL); stock solution and in vivo formulation guidance provided.
  • Storage recommendations: 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.