Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3,5-dibromo-4-hydroxybenzoyl)-2-ethyl-N,N-dimethyl-1-benzofuran-6-sulfonamide | 765317-71-3 | 100.0% | 531.21 | C19H17Br2NO5S | 10 MG
SDP

Catalog No. 5000243081
Encompass_Preferred

PTP1B-IN-15 is a potent, selective small-molecule inhibitor of protein tyrosine phosphatase 1B (PTP1B) for preclinical research into insulin signaling, type II diabetes, and obesity.

  • Potent and selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).
  • Suitable for research into type II diabetes and obesity.
  • High purity (99.95%).
  • Molecular formula C19H17Br2NO5S and molecular weight 531.21.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Soluble in DMSO at 250 mg/mL; may require ultrasonic aid.
  • Powder storage at -20°C; in solvent store at -80°C for long term.

Catalog No. 50-002-43081 Supplier Medchemexpress LLC Supplier No. HY10819610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTP1B-IN-15 is a potent, selective small-molecule inhibitor of protein tyrosine phosphatase 1B (PTP1B) for preclinical research into insulin signaling, type II diabetes, and obesity.

  • Potent and selective inhibitor of protein tyrosine phosphatase 1B (PTP1B).
  • Suitable for research into type II diabetes and obesity.
  • High purity (99.95%).
  • Molecular formula C19H17Br2NO5S and molecular weight 531.21.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Soluble in DMSO at 250 mg/mL; may require ultrasonic aid.
  • Powder storage at -20°C; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.