Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3,5-Dichlorophenoxy)benzaldehyde, 95%, Thermo Scientific™

Catalog No. AC399660010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399660010 Supplier Thermo Scientific Chemicals Supplier No. 399660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81028-92-4
Molecular Formula C13H8Cl2O2
Molecular Weight (g/mol) 267.11
MDL Number MFCD00003355
InChI Key BISWHYILBVQCRA-UHFFFAOYSA-N
Synonym 3-3,5-dichlorophenoxy benzaldehyde, 3-3,5-dichorophenoxy benzaldehyde, m-3,5-dichlorophenoxy benzaldehyde, 3,5-dichloro-3'-formyldiphenyl ether, benzaldehyde,3-3,5-dichlorophenoxy, benzaldehyde, 3-3,5-dichlorophenoxy, pubchem16379, acmc-20anl8, timtec-bb sbb003061, 3-3?5-dichlorophenoxy benzaldehyde
PubChem CID 688050
IUPAC Name 3-(3,5-dichlorophenoxy)benzaldehyde
SMILES ClC1=CC(OC2=CC=CC(C=O)=C2)=CC(Cl)=C1

Specifications

Melting Point 48°C to 55°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula Cl2CH3OC6H4CHO
Quantity 1 g
Formula Weight 267.11
Percent Purity 95%
Chemical Name or Material 3-(3, 5-Dichlorophenoxy)benzaldehyde
Hazard Category GHS Signal Word: Warning
EINECSNumber 279-661-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.