Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3,5-dimethylisoxazol-4-yl)-5-(hydroxy(pyridin-3-yl)methyl)phenol | 2231747-03-6 | MFCD32174354 | 99.0% | 296.32 g·mol⁻¹ | C17H16N2O3 | 5 MG
SDP

Catalog No. 5000222154
Encompass_Preferred

A small-molecule research compound that acts as a potent and selective BRD4 (BET family) inhibitor, with reported cellular activity (IC50 ≈166 nM) and demonstrated antiproliferative effects in cancer cell assays. Supplied as a high-purity solid for biochemical and cellular research applications.

  • Potent BRD4 inhibitor with reported IC50 ≈166 nM.
  • Demonstrated antiproliferative activity in cancer cell assays.
  • Molecular formula C17H16N2O3; molecular weight 296.32 g·mol⁻¹.
  • High purity (98.99%).
  • Supplied as a powder suitable for biochemical and cellular studies.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Catalog No. 50-002-22154 Supplier Medchemexpress LLC Supplier No. HY1352365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule research compound that acts as a potent and selective BRD4 (BET family) inhibitor, with reported cellular activity (IC50 ≈166 nM) and demonstrated antiproliferative effects in cancer cell assays. Supplied as a high-purity solid for biochemical and cellular research applications.

  • Potent BRD4 inhibitor with reported IC50 ≈166 nM.
  • Demonstrated antiproliferative activity in cancer cell assays.
  • Molecular formula C17H16N2O3; molecular weight 296.32 g·mol⁻¹.
  • High purity (98.99%).
  • Supplied as a powder suitable for biochemical and cellular studies.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.