Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(3,5-Dioxo-2,3,4,5-tetrahydro-1,2,4-triazin-6-yl)propanoic acid

Catalog No. OTV0003551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003551G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003551G Supplier Sigma Aldrich Supplier No. OTV0003551G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Linalool oxide is a monoterpenoid compound, commonly found in some species of the aromatic plants
  • It can be obtained from linalool either by oxidation or via biotransfomation using the fungus aspergillus niger
  • It is always present as a mixture of both cis and trans forms

Specifications

CAS 28280-67-3
Molecular Formula C6H7N3O4
Linear Formula C6H7N3O4
MDL Number MFCD00100697
Quantity 25 mL
Molecular Weight (g/mol) 185.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.