Learn More
Cayman Chemical 3,3',5'-Triiodo-L-thyronine, Purity: ≥95%, CAS: 5817-39-0, 10 mg
SDP
Catalog No. 502966629
Encompass_Preferred
A thyroid hormone that is about 1,000-fold less active at the thyroid hormone receptors TRα and TRβ than T3; activates TRΔα1, a native form of TRα that lacks a nuclear localization signal and functions in the cytoplasm, initiating actin polymerization
Catalog No. 50-296-6629 Supplier Cayman Chemical Supplier No. 1759810 MG
A thyroid hormone that is about 1,000-fold less active at the thyroid hormone receptors TRα and TRβ than T3; activates TRΔα1, a native form of TRα that lacks a nuclear localization signal and functions in the cytoplasm, initiating actin polymerization