Learn More
Medchemexpress LLC HY-A0070A 500mg Medchemexpress, 3,3',5-Triiodo-L-thyronine T3;L-3,3',5-Triiodothyronine;Liothyronine;Tresitope CAS:6893-02-3 Purity:98%
SDP
Catalog No. 501639101
Encompass_Preferred
Medchemexpress,HY-A0070A 500mg 3,3',5-Triiodo-L-thyronine T3;L-3,3',5-Triiodothyronine;Liothyronine;Tresitope CAS:6893-02-3 Formula:C15H12I3NO4 3,3',5-Triiodo-L-thyronine is an active form of thyroid hormone, which binds to β1 thyroid hormone receptor (TRβ1), and activates its activity. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9101 Supplier Medchemexpress LLC Supplier No. HYA0070A500MG
Medchemexpress,HY-A0070A 500mg 3,3',5-Triiodo-L-thyronine T3;L-3,3',5-Triiodothyronine;Liothyronine;Tresitope CAS:6893-02-3 Formula:C15H12I3NO4 3,3',5-Triiodo-L-thyronine is an active form of thyroid hormone, which binds to β1 thyroid hormone receptor (TRβ1), and activates its activity. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.