Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5-Triiodo-L-thyronine, 95%, Thermo Scientific Chemicals

Catalog No. AC437260050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437260050 5 g Glass bottle
AC437260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437260050 Supplier Thermo Scientific Chemicals Supplier No. 437260050
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6893-02-3
Molecular Formula C15H12I3NO4
Molecular Weight (g/mol) 650.97
InChI Key AUYYCJSJGJYCDS-UHFFFAOYNA-N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 234°C to 238°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,5565
Specific Rotation +18° to +28° (20°C, 589nm) (c=2, ethanol/0.5N HCl(4:1))
Synonym 4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
SMILES NC(CC1=CC(I)=C(OC2=CC=C(O)C(I)=C2)C(I)=C1)C(O)=O
IUPAC Name 2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
PubChem CID 23359
Formula Weight 650.97
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3', 5-Triiodo-L-thyronine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 229-999-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.