Learn More
Sigma Aldrich Fine Chemicals Biosciences 3,3′,5-Triiodo-L-thyronine sodium salt, 55-06-1, MFCD00002594, 100mg
Catalog No. 501657029
Encompass
Empirical Formula (Hill Notation) C15H11I3NNaO4, Molecular Weight 672.96, powder, BioReagent, suitable for cell culture
Catalog No. 50-165-7029 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T6397100MG
Add to Cart
Add to Cart
Empirical Formula (Hill Notation) C15H11I3NNaO4, Molecular Weight 672.96, powder, BioReagent, suitable for cell culture