Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,5-Trimethylcyclohexyl methacrylate, mixture of isomers, 97%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific™

Catalog No. AAL1219309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1219309 10 g
AAL1219318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1219309 Supplier Thermo Scientific Chemicals Supplier No. L1219309

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7779-31-9
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.317
MDL Number MFCD00078372
InChI Key DABQKEQFLJIRHU-UHFFFAOYSA-N
PubChem CID 111032
IUPAC Name (3,3,5-trimethylcyclohexyl) 2-methylprop-2-enoate
SMILES CC1CC(CC(C1)(C)C)OC(=O)C(=C)C

Specifications

Boiling Point 80°C to 81°C (2 mmHg)
Flash Point 97°C (206°F)
Quantity 10 g
Beilstein 3245062
Synonym 3,3,5-trimethylcyclohexyl methacrylate, 3,5,5-trimethylcyclohexyl methacrylate, 2-propenoic acid, 2-methyl-, 3,3,5-trimethylcyclohexyl ester, 3,3,5-trimethylcyclohexyl 2-methylprop-2-enoate, 3,3,5-trimethylcyclohexyl methacrylate, mixture of isomers, dsstox_cid_24990, dsstox_rid_80636, dsstox_gsid_44990, methacrylic acid 3,3,5-trimethylcyclohexyl ester, 2-propenoic acid,2-methyl-, 3,3,5-trimethylcyclohexyl ester
Formula Weight 210.32
Chemical Name or Material 3,3,5-Trimethylcyclohexyl methacrylate, mixture of isomers
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.