Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,5-Trimethylcyclohexylamine 98.0+%, TCI America™
SDP

Catalog No. T060125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T060125ML Supplier TCI America Supplier No. T060125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15901-42-5
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.26
MDL Number MFCD00019379
InChI Key ZGMQLPDXPUINCQ-UHFFFAOYNA-N
PubChem CID 85931
IUPAC Name 3,3,5-trimethylcyclohexan-1-amine
SMILES CC1CC(N)CC(C)(C)C1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
UN Number 2326
Formula Weight 141.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3,5-Trimethylcyclohexylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.