Learn More
Cayman Chemical 3,3',5-Triiodo-L-thyronine (sodium salt), 55-06-1, 250 mg
SDP
Supplier:  Cayman Chemical 16028250
Encompass_Preferred
A thyroid hormone generated by deiodination of the prohormone thyroxine (T4); a potent agonist of both thyroid hormone receptors TRα and TRβ (Kis = 2.3 nM for both)
Catalog No. NC1382954