Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Bipiperidine 98.0+%, TCI America™
SDP

Catalog No. B43091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43091G Supplier TCI America Supplier No. B43091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 46040-83-9
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.284
InChI Key IKNFMHZKPYNCLN-UHFFFAOYSA-N
PubChem CID 21572583
IUPAC Name 3-piperidin-3-ylpiperidine
SMILES C1CC(CNC1)C2CCCNC2

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bipiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.