Learn More
Medchemexpress LLC [3,3'-Bipyridine]-5-carboxamide, 6'-chloro-N-methyl-5'-[(phenylsulfonyl)amino]- | 2377604-81-2 | 523747 | C18H15ClN4O3S | 25 MG
SDP
Catalog No. 5000374482
Encompass_Preferred
CHMFL-PI4K-127 is an orally active, potent, and highly selective PfPI4K (Plasmodium falciparum PI4K kinase) inhibitor with an IC50 of 0.9 nM. It demonstrates potent activity against 3D7 Plasmodium falciparum, with an EC50 of 25.1 nM, and exhibits antimalaria efficacy.
  • Potent and selective PfPI4K inhibitor.
  • Exhibits antimalaria efficacy.
  • Orally active compound.
Catalog No. 50-003-74482 Supplier Medchemexpress LLC Supplier No. 25MGHY150598
CHMFL-PI4K-127 is an orally active, potent, and highly selective PfPI4K (Plasmodium falciparum PI4K kinase) inhibitor with an IC50 of 0.9 nM. It demonstrates potent activity against 3D7 Plasmodium falciparum, with an EC50 of 25.1 nM, and exhibits antimalaria efficacy.
  • Potent and selective PfPI4K inhibitor.
  • Exhibits antimalaria efficacy.
  • Orally active compound.