Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Bis(bromomethyl)oxetane 95.0+%, TCI America™
SDP

Catalog No. B56411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56411G 1 g
B56415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56411G Supplier TCI America Supplier No. B56411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2402-83-7
Molecular Formula C5H8Br2O
Molecular Weight (g/mol) 243.93
MDL Number MFCD00269694
InChI Key QOPMHMFIIMJWET-UHFFFAOYSA-N
Synonym 2,2-Bis(bromomethyl)-1,3-epoxypropane
PubChem CID 75464
IUPAC Name 3,3-bis(bromomethyl)oxetane
SMILES BrCC1(CBr)COC1

Specifications

Melting Point 25°C
Color White-Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 243.93
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,3-Bis(bromomethyl)oxetane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.