Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Bis(chloromethyl)oxetane

Catalog No. SYX002181G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002181G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002181G Supplier Sigma Aldrich Supplier No. SYX002181G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Perfluorohexyl iodide forms 1:1 complexes with a variety of hydrogen-bond acceptors
  • It has been grafted onto the isocitronellene/propene copolymer by radical reaction yielding a poly(α-olefin) with fluorinated side chains
  • The radical addition of perfluorohexyl iodide to vinyl acetate, using azo-bis( isobutyronitrile) (aibn) as an initiator, has been investigated

Specifications

CAS 78-71-7
Density 1.29 g/mL (at 25°C (literature))
Boiling Point 198°C (lit.)
Molecular Formula C5H8Cl2O
Linear Formula C5H8Cl2O
MDL Number MFCD00027377
Quantity 25 g
Molecular Weight (g/mol) 155.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.