Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Bis[di(p-tolyl)amino]biphenyl 98.0+%, TCI America™
SDP

Catalog No. B48741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48741G 1 g
B48745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48741G Supplier TCI America Supplier No. B48741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161485-60-5
Molecular Formula C40H36N2
Molecular Weight (g/mol) 544.742
MDL Number MFCD16036283
InChI Key AAVPITVHLAHPPZ-UHFFFAOYSA-N
PubChem CID 54123890
IUPAC Name 3-[3-(4-methyl-N-(4-methylphenyl)anilino)phenyl]-N,N-bis(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=CC(=C3)C4=CC(=CC=C4)N(C5=CC=C(C=C5)C)C6=CC=C(C=C6)C

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 1 g
Formula Weight 544.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bis[di(p-tolyl)amino]biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.