Learn More
Echelon Biosciences Research Labs 3,3 -BISOLEOYL-LBPA 500 ug
SDP
Catalog No. 50720540
Encompass
3,3-Bisoleoyl-LBPA ((R,R)-3,3-Bisoleoyl-lysobisphosphatidic acid) is the biologically inactive isomer of LBPA.Lysobisphosphatidic acids (LBPAs), known also as bis-(monoacylglycerol)phosphates (BMPs) are specialized lipids reported to play a role in intracellular protein and lipid transport in healthy cells. Their accumulation was observed in pathologic liver tissue, and they also serve as antigens for auto-antibody generation in a human autoimmune condition termed antiphospholipid syndrome. Accumulation of LBPAs in intracellular, often multilamellar membranes is related to biomembrane polymorphism which may impact intracellular cholesterol transport. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-540 Supplier Echelon Biosciences Research Labs Supplier No. LB182500UG
3,3-Bisoleoyl-LBPA ((R,R)-3,3-Bisoleoyl-lysobisphosphatidic acid) is the biologically inactive isomer of LBPA.Lysobisphosphatidic acids (LBPAs), known also as bis-(monoacylglycerol)phosphates (BMPs) are specialized lipids reported to play a role in intracellular protein and lipid transport in healthy cells. Their accumulation was observed in pathologic liver tissue, and they also serve as antigens for auto-antibody generation in a human autoimmune condition termed antiphospholipid syndrome. Accumulation of LBPAs in intracellular, often multilamellar membranes is related to biomembrane polymorphism which may impact intracellular cholesterol transport. This item is non returnable