Learn More
Sigma Aldrich 3-(3-Bromophenyl)propanenitrile
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- N-hexylmethylamine is an acyclic secondary amine
- Ruthenium-catalyzed reaction of n-hexylmethylamine with styrene has been reported
- Transition metal-catalyzed intermolecular hydroamination of n-hexylmethylamine with 2-vinylnaphthalene has been reported.n-hexylmethylamine may be used in the synthesis of dialkyldithiocarbamato cadmium complexes cd[s2cnrrα]2
Specifications
Specifications
| Molecular Formula | C9H8BrN |
| Linear Formula | C9H8BrN |
| MDL Number | MFCD01310794 |
| Quantity | 10 mL |
| Molecular Weight (g/mol) | 210.07 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.