Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Bromophenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B48655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48655G 5 g
B48651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48655G Supplier TCI America Supplier No. B48655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4422-32-6
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD16658917
InChI Key JHUIVUBQMBPYJE-UHFFFAOYSA-N
PubChem CID 13150401
IUPAC Name 3-(3-bromophenyl)pyridine
SMILES C1=CC(=CC(=C1)Br)C2=CN=CC=C2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(3-Bromophenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.