Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Carbonylbis(7-diethylaminocoumarin), 99%

Catalog No. ac404050050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404050050 Supplier Thermo Scientific Chemicals Supplier No. 404050050
Only null left
Add to Cart

CAS: 63226-13-1 | C27H28N2O5 | 460.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63226-13-1
Molecular Formula C27H28N2O5
Molecular Weight (g/mol) 460.52
MDL Number MFCD00050047
InChI Key SANIRTQDABNCHF-UHFFFAOYSA-N
Synonym 3,3'-carbonylbis 7-diethylaminocoumarin, 3,3'-carbonylbis 7-diethylamino-2h-chromen-2-one, 2h-1-benzopyran-2-one, 3,3'-carbonylbis 7-diethylamino, 7-diethylamino-3-7-diethylamino-2-oxochromene-3-carbonyl chromen-2-one, 7-diethylamino-3-7-diethylamino-2-oxochromen-3-yl carbonyl chromen-2-one, 3,3'-carbonyl-bis-7-diethylaminocoumarin, 2h-1-benzopyran-2-one, 3,3-carbonylbis 7-diethylamino, 7-diethylamino-3-7-diethylamino-2-oxo-2h-chromen-3-yl carbonyl-2h-ch romen-2-one
PubChem CID 113160
IUPAC Name 7-(diethylamino)-3-[7-(diethylamino)-2-oxochromene-3-carbonyl]chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)C3=CC4=C(C=C(C=C4)N(CC)CC)OC3=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 214.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 460.52
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Carbonylbis(7-diethylaminocoumarin), 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.