Learn More
Selleck Chemical LLC 3 3 -cGAMP
SDP
Catalog No. 5000000051
Encompass_Preferred
3 3 -cGAMP (3 3 -cyclic GMP-AMP Cyclic GMP-AMP cGAMP) activates the endoplasmic reticulum (ER)-resident receptor stimulator of interferon genes (STING) thereby inducing an antiviral state and the secretion of type I IFNs
Catalog No. 50-000-00051 Supplier Selleck Chemical LLC Supplier No. S79051MG
3 3 -cGAMP (3 3 -cyclic GMP-AMP Cyclic GMP-AMP cGAMP) activates the endoplasmic reticulum (ER)-resident receptor stimulator of interferon genes (STING) thereby inducing an antiviral state and the secretion of type I IFNs