Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3-chlorophenyl)-6-(trifluoromethyl)-1H-benzimidazol-2-one | 625458-06-2 | 99.9% | 5 MG
SDP

Catalog No. 5000233079
Encompass_Preferred

UCCF-853 is a small-molecule CFTR modulator for research use (CAS 625458-06-2). It is provided as a white to off-white solid with reported high purity, intended for in vitro and preclinical research applications. Offered in small milligram quantities.

  • Small-molecule CFTR modulator for research use.
  • CAS 625458-06-2.
  • Molecular formula C14H8ClF3N2O; molecular weight 312.67 g/mol.
  • White to off-white solid appearance.
  • High reported purity (99.9%).
  • Available in small research sizes such as 5 MG and 10 MG.

Catalog No. 50-002-33079 Supplier Medchemexpress LLC Supplier No. HY1191795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UCCF-853 is a small-molecule CFTR modulator for research use (CAS 625458-06-2). It is provided as a white to off-white solid with reported high purity, intended for in vitro and preclinical research applications. Offered in small milligram quantities.

  • Small-molecule CFTR modulator for research use.
  • CAS 625458-06-2.
  • Molecular formula C14H8ClF3N2O; molecular weight 312.67 g/mol.
  • White to off-white solid appearance.
  • High reported purity (99.9%).
  • Available in small research sizes such as 5 MG and 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.