Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl 98.0+%, TCI America™
SDP

Catalog No. D4772200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4772200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4772200MG Supplier TCI America Supplier No. D4772200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 342638-54-4
Molecular Formula C36H24N2
Molecular Weight (g/mol) 484.602
MDL Number MFCD28138084
InChI Key NSXJEEMTGWMJPY-UHFFFAOYSA-N
Synonym mCBP
PubChem CID 23386664
IUPAC Name 9-[3-(3-carbazol-9-ylphenyl)phenyl]carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=CC(=C4)C5=CC(=CC=C5)N6C7=CC=CC=C7C8=CC=CC=C86

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 200 mg
Formula Weight 484.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Di(9H-carbazol-9-yl)-1,1′-biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.