Learn More
Medchemexpress LLC 3,3'-Di-O-methylellagic acid | 2239-88-5 | 330.25 | 5 MG
SDP
Catalog No. 5000416516
Encompass_Preferred
3,3'-Di-O-methylellagic acid is a compound obtained from *Euphorbia adenochlora*. It selectively inhibits the formation of acid-fastness in mycobacteria without retarding their growth and also acts as a hepatoprotective compound due to its antioxidative effect.
  • Selectively inhibits acid-fastness in mycobacteria.
  • Does not retard mycobacteria growth.
  • Acts as a hepatoprotective compound.
  • Exhibits antioxidative effects.
  • Demonstrates anticomplement activity in erythrocytes.
Catalog No. 50-004-16516 Supplier Medchemexpress LLC Supplier No. HYN19695MG
3,3'-Di-O-methylellagic acid is a compound obtained from *Euphorbia adenochlora*. It selectively inhibits the formation of acid-fastness in mycobacteria without retarding their growth and also acts as a hepatoprotective compound due to its antioxidative effect.
  • Selectively inhibits acid-fastness in mycobacteria.
  • Does not retard mycobacteria growth.
  • Acts as a hepatoprotective compound.
  • Exhibits antioxidative effects.
  • Demonstrates anticomplement activity in erythrocytes.