Learn More
Medchemexpress LLC 3,3'-Di-O-methylellagic acid | 2239-88-5 | MFCD01083342 | 330.25 | 1 MG
SDP
Catalog No. 5000414810
Encompass_Preferred
3,3'-Di-O-methylellagic acid, also known as 3,8-Di-O-methylellagic acid, is a chemical compound obtained from *Euphorbia adenochlora*. It selectively inhibits the formation of acid-fastness in mycobacteria without hindering their growth. Additionally, it exhibits hepatoprotective effects, which are attributed to its antioxidative properties.
  • Inhibits the formation of acid-fastness in mycobacteria.
  • Does not retard the growth of mycobacteria.
  • Acts as a hepatoprotective compound.
  • Exhibits antioxidative effects.
Catalog No. 50-004-14810 Supplier Medchemexpress LLC Supplier No. HYN19691MG
3,3'-Di-O-methylellagic acid, also known as 3,8-Di-O-methylellagic acid, is a chemical compound obtained from *Euphorbia adenochlora*. It selectively inhibits the formation of acid-fastness in mycobacteria without hindering their growth. Additionally, it exhibits hepatoprotective effects, which are attributed to its antioxidative properties.
  • Inhibits the formation of acid-fastness in mycobacteria.
  • Does not retard the growth of mycobacteria.
  • Acts as a hepatoprotective compound.
  • Exhibits antioxidative effects.