Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,3'-Diaminobenzidine, 99%

Manufacturer:  Alfa Aesar™ H5400022

Quantity 100g

 View more versions of this product

Catalog No. AAH5400022


  Check Availability
Add to cart

Specifications

Description

Applications
3,3 ft.-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

Solubility
Soluble in water (partly).

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
Description & Specifications

Chemical Identifiers

91-95-2
214.27
DAB; 3,3',4,4'-Tetraaminobiphenyl
C12H14N4
MFCD00007725
3,3'-Diaminobenzidine

Specifications

175° to 177°C
91-95-2
214.27
DAB; 3,3',4,4'-Tetraaminobiphenyl
99%
100g
C12H14N4
MFCD00007725
3,3'-Diaminobenzidine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H341
May cause cancer.
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DV8750000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit