Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diaminobenzidine Tetrahydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. D37571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37571G 1 g
D37575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37571G Supplier TCI America Supplier No. D37571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 868272-85-9
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
PubChem CID 23191111
IUPAC Name [1,1'-biphenyl]-3,3',4,4'-tetramine tetrahydrochloride
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1

Specifications

Melting Point 280°C
Color Purple-White
Quantity 1 g
Formula Weight 360.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diaminobenzidine Tetrahydrochloride Hydrate [for Biochemical Research]
RTECSNumber DV8753000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.