Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diaminobenzidine Tetrahydrochloride Hydrate, Sigma-Aldrich

Catalog No. 01258842 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-842 5 g
01-258-971 50 g
01-259-015 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-258-842 Supplier MilliporeSigma Supelco Supplier No. D56375G
Only null left
Add to Cart
Add to Cart

In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidized DAB forms an insoluble brown-colored complex.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stain
Quantity 5 g
For Use With (Application) Immunohistochemistry (IHC)
Concentration ≥96%
Form Powder, crystals, or chunks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.