Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diaminobenzidine Tetrahydrochloride Hydrate, Sigma-Aldrich

Catalog No. 01259015 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-015 10 g
01-258-842 5 g
01-258-971 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-259-015 Supplier MilliporeSigma Supelco Supplier No. D563710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidized DAB forms an insoluble brown-colored complex.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stain
Quantity 10 g
For Use With (Application) Immunohistochemistry (IHC)
Concentration ≥96%
Form Powder, crystals, or chunks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.