Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diaminobenzophenone, Thermo Scientific™

Catalog No. AA3121903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3121903 1 g
AA3121906 5 g
AA3121914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3121903 Supplier Thermo Scientific Chemicals Supplier No. 3121903

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-79-0
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00014774
InChI Key TUQQUUXMCKXGDI-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
PubChem CID 69145
IUPAC Name 3-(3-aminobenzoyl)aniline
SMILES NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 150°C to 151°C
Boiling Point 285°C (11 mmHg)
Quantity 1 g
Formula Weight 212.25
Physical Form Powder
Chemical Name or Material 3,3'-Diaminobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.