Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dichloro-2-formoxytetrahydropyran 98+%, Thermo Scientific™

Catalog No. AC278480025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC278480025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC278480025 Supplier Thermo Scientific Chemicals Supplier No. 278480025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141942-53-2
Molecular Formula C6H8Cl2O3
Molecular Weight (g/mol) 199.027
InChI Key RJZSBBQFYWZFDG-UHFFFAOYSA-N
Synonym 3,3-dichlorooxan-2-yl formate, 3,3-dichloro-2-formoxytetrahydropyran, 2h-pyran-2-ol,3,3-dichlorotetrahydro-, 2-formate, 3,3-dichloro-2h-3,4,5,6-tetrahydropyran-2-yl formate, acmc-20n10x, 3,3-dichlorotetrahydro-2h-pyran-2-yl formate
PubChem CID 2724838
IUPAC Name (3,3-dichlorooxan-2-yl) formate
SMILES C1CC(C(OC1)OC=O)(Cl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4835 to 1.4855 (20°C, 589nm)
Quantity 2.5 g
Formula Weight 199.03
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 3, 3-Dichloro-2-formoxytetrahydropyran, 98+%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.