Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dichlorobenzidine, MilliporeSigma™ Supelco™

Catalog No. 111019380 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9380 Supplier MilliporeSigma Supelco Supplier No. 48525
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 91-94-1
Physical Form Neat
Linear Formula C12H10 Cl2N2
Type Food and Cosmetic Component Standard
Quantity 100 mg
Formula Weight 253.13
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.