Learn More
eMolecules​ 16777-87-0 | 3,3-DIETHOXY-1-PROPANOL | AstaTech | MFCD00074850 | 148.202 | C7H16O3 | 98.000 | CCOC(CCO)OCC | 25g | 248475942
Catalog No. NC2225818 Shop All eMolecules Inc. Products
Encompass_Preferred
3,3-DIETHOXY-1-PROPANOL | AstaTech | 16777-87-0 | MFCD00074850 | 148.202 | C7H16O3 | 98.000 | CCOC(CCO)OCC | 25g | 248475942
Catalog No. NC2225818 Supplier eMolecules​ Supplier No. 248475942
3,3-DIETHOXY-1-PROPANOL | AstaTech | 16777-87-0 | MFCD00074850 | 148.202 | C7H16O3 | 98.000 | CCOC(CCO)OCC | 25g | 248475942