Learn More
Cayman Chemical 3 3-DIethyloxacarbocynIn 10mg
SDP
Catalog No. 502916952
Encompass_Preferred
A fluorescent dye; has been used to monitor mammalian and bacterial membrane potential; upon cell hyperpolarization, it enters cells, and exhibits a decrease in fluorescence intensity and shift in emission from 530 nm to 670 nm upon excitation at 488 nm; selectively inhibits bovine heart mitochondrial complex I (IC50 = 9 µM) over bovine heart mitochondrial complex IV and P. denitrificans mitochondrial complex I at 10 µM; activates bovine heart mitochondrial complex II at 10 µM
Catalog No. 50-291-6952 Supplier Cayman Chemical Supplier No. 3945610 MG
A fluorescent dye; has been used to monitor mammalian and bacterial membrane potential; upon cell hyperpolarization, it enters cells, and exhibits a decrease in fluorescence intensity and shift in emission from 530 nm to 670 nm upon excitation at 488 nm; selectively inhibits bovine heart mitochondrial complex I (IC50 = 9 µM) over bovine heart mitochondrial complex IV and P. denitrificans mitochondrial complex I at 10 µM; activates bovine heart mitochondrial complex II at 10 µM