Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,3-Difluoropyrrolidine hydrochloride | 163457-23-6 | 143.56 | 25 G
SDP

Catalog No. 5000449894
Encompass_Preferred

3,3-Difluoropyrrolidine hydrochloride is a biochemical reagent suitable for life science related research. It can be utilized as a biological material or an organic compound.

  • Solid appearance
  • White to off-white color
  • Purity of 99.5%
  • Molecular weight of 143.56
  • Chemical formula C4H8ClF2N
  • CAS number 163457-23-6

Catalog No. 50-004-49894 Supplier Medchemexpress LLC Supplier No. 25GHYW001466
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,3-Difluoropyrrolidine hydrochloride is a biochemical reagent suitable for life science related research. It can be utilized as a biological material or an organic compound.

  • Solid appearance
  • White to off-white color
  • Purity of 99.5%
  • Molecular weight of 143.56
  • Chemical formula C4H8ClF2N
  • CAS number 163457-23-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.