Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,3-Difluoropyrrolidine hydrochloride | 163457-23-6 | 143.56 | 50 G
SDP

Catalog No. 5000447246
Encompass_Preferred

3,3-Difluoropyrrolidine hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a white to off-white solid.

  • Biochemical reagent
  • Used as a biological material or organic compound for life science related research
  • For research use only
  • White to off-white solid appearance
  • Assay: 99.5%
  • Molecular weight: 143.56
  • Storage at 4°C, sealed, away from moisture
  • In solvent storage at -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-47246 Supplier Medchemexpress LLC Supplier No. HYW00146650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,3-Difluoropyrrolidine hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a white to off-white solid.

  • Biochemical reagent
  • Used as a biological material or organic compound for life science related research
  • For research use only
  • White to off-white solid appearance
  • Assay: 99.5%
  • Molecular weight: 143.56
  • Storage at 4°C, sealed, away from moisture
  • In solvent storage at -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.