Learn More
Medchemexpress LLC HY-D0084 50mg Medchemexpress, 3,3'-Dihexyloxacarbocyanine iodide CAS:53213-82-4 Purity:>98%
SDP
Catalog No. 501933147
Encompass_Preferred
Medchemexpress, HY-D0084 50mg 3,3'-Dihexyloxacarbocyanine iodide CAS:53213-82-4 3,3'-Dihexyloxacarbocyanine iodide is a carbocyanine dye which can be used to monitor changes in mitochondrial membrane potential. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3147 Supplier Medchemexpress LLC Supplier No. HYD008450MG
Medchemexpress, HY-D0084 50mg 3,3'-Dihexyloxacarbocyanine iodide CAS:53213-82-4 3,3'-Dihexyloxacarbocyanine iodide is a carbocyanine dye which can be used to monitor changes in mitochondrial membrane potential. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.