Learn More
Enzo Life Sciences 3,3'-Diindolylmethane (500mg). CAS: 1968-05-4
SDP
Catalog No. 502008867
Encompass_Preferred
Dimer of indole-3-carbinol (ALX-350-347). Anticancer and antineoplastic agent. Induces apoptosis in human cancer cells. Activator of cytochrome P450. Partial agonist of aryl hydrocarbon receptor (AhR). Alternative name: DIM. Purity: ≥98% (HPLC). Appearance: White to light pink powder. Solubility: Soluble in DMSO (200mg/ml); insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8867 Supplier Enzo Life Sciences Supplier No. ALX350370M500
Dimer of indole-3-carbinol (ALX-350-347). Anticancer and antineoplastic agent. Induces apoptosis in human cancer cells. Activator of cytochrome P450. Partial agonist of aryl hydrocarbon receptor (AhR). Alternative name: DIM. Purity: ≥98% (HPLC). Appearance: White to light pink powder. Solubility: Soluble in DMSO (200mg/ml); insoluble in water. Long Term Storage: -20°C.