Learn More
Selleck Chemical LLC 3.3-Diindolylmethane S4728-100mg
SDP
Catalog No. 503108434
Encompass_Preferred
3 3 -Diindolylmethane(DIM) is a major digestive product of indole-3-carbinol a potential anticancer component of cruciferous vegetables 3 3 -Diindolylmethane is a strong pure androgen receptor (AR) antagonist
Catalog No. 50-310-8434 Supplier Selleck Chemical LLC Supplier No. S4728100MG
3 3 -Diindolylmethane(DIM) is a major digestive product of indole-3-carbinol a potential anticancer component of cruciferous vegetables 3 3 -Diindolylmethane is a strong pure androgen receptor (AR) antagonist