Learn More
Selleck Chemical LLC 3 3-Dimethyl-1-butanol E4432-100MG
SDP
Catalog No. 5000039515
Encompass_Preferred
3 3-Dimethyl-1-butanol (Neohexanol) is an orally active inhibitor of trimethylamine (TMA) and trimethylamine N-oxide (TMAO) 3 3-dimethyl-1-butanol negatively inhibits the signalling pathways of p65 NF- B and TGF- 1/Smad3 3 3-dimethyl-1-butanol has potential applications in cardiovascular disease (CVD)
Catalog No. 50-000-39515 Supplier Selleck Chemical LLC Supplier No. E4432100MG
3 3-Dimethyl-1-butanol (Neohexanol) is an orally active inhibitor of trimethylamine (TMA) and trimethylamine N-oxide (TMAO) 3 3-dimethyl-1-butanol negatively inhibits the signalling pathways of p65 NF- B and TGF- 1/Smad3 3 3-dimethyl-1-butanol has potential applications in cardiovascular disease (CVD)