Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-indanone 95.0+%, TCI America™
SDP

Catalog No. D31605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31605G 5 g
D31601G 1 g
D316025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D31605G Supplier TCI America Supplier No. D31605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26465-81-6
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD01846169
InChI Key QWZAOSKLFKAEOK-UHFFFAOYSA-N
PubChem CID 304628
IUPAC Name 3,3-dimethyl-2H-inden-1-one
SMILES CC1(CC(=O)C2=CC=CC=C21)C

Specifications

Boiling Point 122°C
Quantity 5 g
Formula Weight 160.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-indanone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.