Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-2-butanol, 98+%, Thermo Scientific Chemicals

Catalog No. AA4372550
Change view
Click to view available options
Quantity:
5 x 10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4372550 5 x 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4372550 Supplier Thermo Scientific Chemicals Supplier No. 04372550

missing translation for 'validMainframeMsg'

3,3-Dimethyl-2-butanol was used in conversion of ribose- and glucose-binding proteins into receptors for pinacolyl methyl phosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-07-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004522
InChI Key DFOXKPDFWGNLJU-UHFFFAOYSA-N
Synonym 3,3-dimethyl-2-butanol, pinacolyl alcohol, 2-butanol, 3,3-dimethyl, 2,2-dimethyl-3-butanol, tert-butyl methyl carbinol, 3,3-dimethylbutane-2-ol, 2-butanol, 3,3-dimethyl-, s, s-3,3-dimethyl-2-butanol, pinacolone alcohol, 3,3-dimethyl 2-butanol
PubChem CID 10045
IUPAC Name 3,3-dimethylbutan-2-ol
SMILES CC(C(C)(C)C)O

Specifications

Melting Point 5.3°C
Density 0.812 g/mL
Boiling Point 119°C to 121°C
Flash Point 28°C (84°F)
Refractive Index 1.415
Quantity 5 x 10 g
UN Number UN1987
Solubility Information Soluble in ethanol,diethyl ether,water (25g/L).
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-butanol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 207-347-9
RTECSNumber EL2276000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.