Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Dimethyl-2-butanone

Catalog No. P45605500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P45605500ML 500 mL
P45605100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P45605500ML Supplier Sigma Aldrich Supplier No. P45605500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,3-Dimethyl-2-butanone is an aliphatic ketone can undergo asymmetric reduction to the corresponding alcohol with diisopinocampheylchloroborane with high enantiomeric excess.

Specifications

CAS 75-97-8
Density 0.801 g/mL (at 25°C (literature))
Boiling Point 106°C (lit.)
Molecular Formula C6H12O
Refractive Index n20/D 1.396 (literature)
Linear Formula CH3COC(CH3)3
MDL Number MFCD00008846
Quantity 500 mL
Synonym alpha,alpha,alpha-Trimethylacetone; tert-Butyl methyl ketone; Pinacolone
Molecular Weight (g/mol) 100.16
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.