Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-dimethyl-4-nitrobutanoic acid

Catalog No. VBP00283 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP00283 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP00283 Supplier Sigma Aldrich Supplier No. VBP00283500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tributyl o-acetylcitrate (tboac) can be used:
  • As a plasticizer to improve flexibility and impact properties of polylactide (pla) polymer
  • As a processing additive for the formulation of bulk heterojunction (bhj) polymeric organic solar cells (oscs) to improve their efficiency
  • In the preparation of semiconducting biopolymer composites of poly(3-hydroxy butyrate) (phb)
  • In the synthesis of functionalized poly(vinyl chloride)(pvc) membranes for selective separation of perchlorate from water

Specifications

Molecular Formula C6H11O4N1
Linear Formula C6H11O4N1
MDL Number MFCD30471719
Quantity 500 mL
Molecular Weight (g/mol) 161.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.