Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Dimethylacrylic acid

Catalog No. D138606500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D138606500G 500 g
D138606100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D138606500G Supplier Sigma Aldrich Supplier No. D138606500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,3-Dimethylacrylic acid

Specifications

CAS 541-47-9
Melting Point 65°C to 70°C (lit.)
Boiling Point 194°C to 195°C (lit.)
Molecular Formula C5H8O2
Linear Formula (CH3)2 C=CHCOOH
MDL Number MFCD00004366
Quantity 500 g
Synonym 3-Methyl-2-butenoic acid; 3,3-Dimethylacrylic acid; 3-Methylcrotonic acid; Senecioic acid
Molecular Weight (g/mol) 100.12
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.