Learn More
Medchemexpress LLC 3,3-Dimethylacrylic acid | 541-47-9 | 99.9% | 100.12 | 25 G
SDP
Catalog No. 5000451651
Encompass_Preferred
3,3-Dimethylacrylic acid is an active small molecule that can be used as a drug intermediate. It is for research use only. It exhibits antituberculosis activity with a MIC of 0.078 μg/mL and can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid.
  • Used as a drug intermediate.
  • Exhibits antituberculosis activity with a MIC of 0.078 μg/mL.
  • Can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid.
  • Functions as a human endogenous metabolite.
  • For research use only.
Catalog No. 50-004-51651 Supplier Medchemexpress LLC Supplier No. 25GHYW010611
Add to Cart
Add to Cart
3,3-Dimethylacrylic acid is an active small molecule that can be used as a drug intermediate. It is for research use only. It exhibits antituberculosis activity with a MIC of 0.078 μg/mL and can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid.
  • Used as a drug intermediate.
  • Exhibits antituberculosis activity with a MIC of 0.078 μg/mL.
  • Can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid.
  • Functions as a human endogenous metabolite.
  • For research use only.