Learn More
Medchemexpress LLC 3,3-Dimethylacrylic acid | 541-47-9 | 99.99% | 100.12 | 100 G
SDP
Catalog No. 5000366553
Encompass_Preferred
3,3-Dimethylacrylic acid is an active small molecule that functions as a drug intermediate. It exhibits antituberculosis activity with a MIC of 0.078 μg/mL and can be utilized in the synthesis of triorganotin(IV) complexes of cinnamic acid. This compound is also recognized as an endogenous metabolite.
  • Active small molecule
  • Drug intermediate
  • Exhibits antituberculosis activity
  • Used for synthesis of triorganotin(IV) complexes of cinnamic acid
  • Endogenous metabolite
Catalog No. 50-003-66553 Supplier Medchemexpress LLC Supplier No. HYW010611100G
3,3-Dimethylacrylic acid is an active small molecule that functions as a drug intermediate. It exhibits antituberculosis activity with a MIC of 0.078 μg/mL and can be utilized in the synthesis of triorganotin(IV) complexes of cinnamic acid. This compound is also recognized as an endogenous metabolite.
  • Active small molecule
  • Drug intermediate
  • Exhibits antituberculosis activity
  • Used for synthesis of triorganotin(IV) complexes of cinnamic acid
  • Endogenous metabolite